Anti-DDX3X Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1660-22EA 3340 DKK
USBID1660-22
Anti-DDX3X Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: DDX3X (DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR