Anti-DDX3X Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1660-20E-APCEA 6360 DKK
USBID1660-20E-APC
Anti-DDX3X Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: DDX3X (DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR