Anti-DDX3X Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1660-20CEA 4140 DKK
USBID1660-20C
Anti-DDX3X Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: DDX3X (DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR