Anti-DDX1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2679]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1660-17C-HRPEA 0 DKK
USBID1660-17C-HRP
Anti-DDX1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2679]
Type: Primary
Antigen: DDX1 (DEAD (Asp-Glu-Ala-Asp) box helicase 1)
Clonality: monoclonal
Clone: 10B2679
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR