Anti-DDR1 Rabbit monoclonal antibody [clone: 11C1024]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1649-03GEA 3610 DKK
USBID1649-03G
Anti-DDR1 Rabbit monoclonal antibody [clone: 11C1024]
Antistoffer
Recommended dilution: Immunofluorescence: 1:800; Western Blot: 1:1000; Immunoprecipitation: 1:100; Immunochemistry: 1:6400

Type: Primary
Antigen: DDR1 (discoidin domain receptor tyrosine kinase 1)
Clonality: monoclonal
Clone: 11C1024
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR