Anti-DDR1 Mouse monoclonal antibody [clone: 2G4E12]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1649-01GEA 4320 DKK
USBID1649-01G
Anti-DDR1 Mouse monoclonal antibody [clone: 2G4E12]
Antistoffer
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:300

Type: Primary
Antigen: DDR1 (discoidin domain receptor tyrosine kinase 1)
Clonality: monoclonal
Clone: 2G4E12
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR