Anti-DCAF6 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1575-30EA 3180 DKK
USBID1575-30
Anti-DCAF6 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1-3?g/ml

Type: Primary
Antigen: DCAF6 (DDB1 and CUL4 associated factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR