Anti-DDB1 Mouse monoclonal antibody [clone: 1322]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1575-01HEA 4320 DKK
USBID1575-01H
Anti-DDB1 Mouse monoclonal antibody [clone: 1322]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: monoclonal
Clone: 1322
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR