Anti-DDB1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1575-01GEA 4320 DKK
USBID1575-01G
Anti-DDB1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR