Anti-DDB1 Mouse monoclonal antibody [clone: 6A63]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1575-01AEA 3980 DKK
USBID1575-01A
Anti-DDB1 Mouse monoclonal antibody [clone: 6A63]
Antistoffer
Recommended dilution: Immunoprecipitation: 5-10 ?g/reaction; Western Blot: 1-5 ?g/ml; ELISA: 0.1-1 ?g/ml

Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: monoclonal
Clone: 6A63
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR