Anti-DDAH2 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1560-01C1EA 3460 DKK
USBID1560-01C1
Anti-DDAH2 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:2000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: DDAH2 (dimethylarginine dimethylaminohydrolase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR