Anti-DDAH1 Goat polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID1559-95DEA 4120 DKK
USBID1559-95D
Anti-DDAH1 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:16000; Western Blot: 0.1-0. 3?g/ml
Type: Primary
Antigen: DDAH1 (dimethylarginine dimethylaminohydrolase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human