Anti-DCXR Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1550-50DEA 4120 DKK
USBID1550-50D
Anti-DCXR Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: DCXR (dicarbonyl/L-xylulose reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR