Anti-CD209 Mouse Monoclonal Antibody (Biotin) [clone: 9G179]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1550-02BEA 4250 DKK
USBID1550-02B
Anti-CD209 Mouse Monoclonal Antibody (Biotin) [clone: 9G179]
Antistoffer
Type: Primary
Antigen: CD209 (CD209 molecule)
Clonality: monoclonal
Clone: 9G179
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR