Anti-CD209 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B1042]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1550-01D5EA 4730 DKK
USBID1550-01D5
Anti-CD209 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B1042]
Antistoffer
Recommended dilution: Flow Cytometry:1:5;

Type: Primary
Antigen: CD209 (CD209 molecule)
Clonality: monoclonal
Clone: 10B1042
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR