Anti-TNFRSF6B Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2676]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1500-25R-PEEA 0 DKK
USBID1500-25R-PE
Anti-TNFRSF6B Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2676]
Type: Primary
Antigen: TNFRSF6B (tumor necrosis factor receptor superfamily, member 6b, decoy)
Clonality: monoclonal
Clone: 10B2676
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR