Anti-TNFRSF6B Mouse Monoclonal Antibody (Biotin) [clone: 9i230]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID1500-25LEA 6740 DKK
USBID1500-25L
Anti-TNFRSF6B Mouse Monoclonal Antibody (Biotin) [clone: 9i230]
Antistoffer
Recommended dilution: ELISA: 0.5-2?g/ml
Type: Primary
Antigen: TNFRSF6B (tumor necrosis factor receptor superfamily, member 6b, decoy)
Clonality: monoclonal
Clone: 9i230
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human