Anti-TNFRSF6B Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1500-25B2EA 3350 DKK
USBID1500-25B2
Anti-TNFRSF6B Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: TNFRSF6B (tumor necrosis factor receptor superfamily, member 6b, decoy)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR