Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 6D346]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1499-04EA 4100 DKK
USBID1499-04
Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 6D346]
Antistoffer
Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: monoclonal
Clone: 6D346
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR