Anti-TNFRSF10C Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1499-03AEA 2990 DKK
USBID1499-03A
Anti-TNFRSF10C Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR