Anti-DCUN1D1 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1493-55EA 3460 DKK
USBID1493-55
Anti-DCUN1D1 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:16. 000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: DCUN1D1 (DCN1, defective in cullin neddylation 1, domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR