Anti-DCTD Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2673]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1490-02-FITCEA 0 DKK
USBID1490-02-FITC
Anti-DCTD Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2673]
Type: Primary
Antigen: DCTD (dCMP deaminase)
Clonality: monoclonal
Clone: 10B2673
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR