Anti-DCTD Chicken polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1490-01EA 3240 DKK
USBID1490-01
Anti-DCTD Chicken polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: DCTD (dCMP deaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR