Anti-DCC Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1200-01BEA 3880 DKK
USBID1200-01B
Anti-DCC Goat polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 2.5-5 ?g/ml

Type: Primary
Antigen: DCC (deleted in colorectal carcinoma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR