Anti-DCAKD Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2672]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1198-85-HRPEA 0 DKK
USBID1198-85-HRP
Anti-DCAKD Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2672]
Type: Primary
Antigen: DCAKD (dephospho-CoA kinase domain containing)
Clonality: monoclonal
Clone: 10B2672
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR