Anti-DBC-1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 11C280]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1171-01E-PEEA 0 DKK
USBID1171-01E-PE
Anti-DBC-1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 11C280]
Type: Primary
Antigen: DBC-1
Clonality: monoclonal
Clone: 11C280
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR