Anti-DAXX Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1150-04EA 4730 DKK
USBID1150-04
Anti-DAXX Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:7000-1:45000; Western Blot: 1:200-1:2000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: DAXX (death-domain associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR