Anti-DARC Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1101-01EA 3340 DKK
USBID1101-01
Anti-DARC Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR