Anti-DIO1 Rabbit Polyclonal Antibody (Biotin)
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID1080-15-BIOTEA 6360 DKK
USBID1080-15-BIOT
Anti-DIO1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DIO1 (deiodinase, iodothyronine, type I)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human