Anti-DARPP32 Rabbit monoclonal antibody [clone: 7H17]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1075-07EA 3350 DKK
USBID1075-07
Anti-DARPP32 Rabbit monoclonal antibody [clone: 7H17]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50; Immunochemistry: 1:200

Type: Primary
Antigen: DARPP32
Clonality: monoclonal
Clone: 7H17
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR