Anti-DARPP32 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1075-06EA 4750 DKK
USBID1075-06
Anti-DARPP32 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: DARPP32
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR