Anti-DARPP32 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1075-05BEA 2590 DKK
USBID1075-05B
Anti-DARPP32 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: DARPP32
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR