Anti-DARPP32 Rabbit monoclonal antibody [clone: 11C1022]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1075-03DEA 2900 DKK
USBID1075-03D
Anti-DARPP32 Rabbit monoclonal antibody [clone: 11C1022]
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: DARPP32
Clonality: monoclonal
Clone: 11C1022
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR