Anti-DARC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H31]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID1065-50CEA 5300 DKK
USBID1065-50C
Anti-DARC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H31]
Antistoffer
Do not freeze!
Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: monoclonal
Clone: 7H31
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human