Anti-DARC Mouse monoclonal antibody [clone: 7H31]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1065-50BEA 4320 DKK
USBID1065-50B
Anti-DARC Mouse monoclonal antibody [clone: 7H31]
Antistoffer
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: monoclonal
Clone: 7H31
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR