Anti-DARC Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1065-50EA 3340 DKK
USBID1065-50
Anti-DARC Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR