Anti-DACT1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1055-02AEA 3180 DKK
USBID1055-02A
Anti-DACT1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: DACT1 (dishevelled-binding antagonist of beta-catenin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat, Xenopus laevis, Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR