Anti-DAPK2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1052-01D-BIOTEA 4620 DKK
USBID1052-01D-BIOT
Anti-DAPK2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DAPK2 (death-associated protein kinase 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR