Anti-DAP Kinase 2 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1052-01AEA 3460 DKK
USBID1052-01A
Anti-DAP Kinase 2 Goat polyclonal antibody
Antistoffer
Type: Primary
Antigen: DAP Kinase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR