Anti-DAPK1 Rabbit polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID1051-02AEA 4180 DKK
USBID1051-02A
Anti-DAPK1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: DAPK1 (death-associated protein kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human