Anti-TYROBP Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1049-06EA 2940 DKK
USBID1049-06
Anti-TYROBP Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: TYROBP (TYRO protein tyrosine kinase binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR