Anti-DAP3 Mouse monoclonal antibody [clone: 8J1260]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1049-03EEA 3180 DKK
USBID1049-03E
Anti-DAP3 Mouse monoclonal antibody [clone: 8J1260]
Antistoffer
Type: Primary
Antigen: DAP3 (death associated protein 3)
Clonality: monoclonal
Clone: 8J1260
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR