Anti-DAB1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID0900-15EA 3350 DKK
USBID0900-15
Anti-DAB1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: DAB1 (Dab, reelin signal transducer, homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR