Anti-D4-GDI Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11E166]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID0100-01CEA 3970 DKK
USBID0100-01C
Anti-D4-GDI Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11E166]
Antistoffer
Type: Primary
Antigen: D4-GDI
Clonality: monoclonal
Clone: 1.1E+167
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR