Anti-Cytomegalovirus Mouse monoclonal antibody [clone: 8A269]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9100-23CEA 3760 DKK
USBIC9100-23C
Anti-Cytomegalovirus Mouse monoclonal antibody [clone: 8A269]
Antistoffer
Type: Primary
Antigen: Cytomegalovirus
Clonality: Monoclonal
Clone: 8A269
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR