Anti-Cytomegalovirus Glycoprotein H, Late Neutralizing Antigen Mouse monoclonal antibody [clone: 10B2667]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9100-22FEA 4060 DKK
USBIC9100-22F
Anti-Cytomegalovirus Glycoprotein H, Late Neutralizing Antigen Mouse monoclonal antibody [clone: 10B2667]
Antistoffer
Type: Primary
Antigen: Cytomegalovirus Glycoprotein H, Late Neutralizing Antigen
Clonality: Monoclonal
Clone: 10B2667
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR