Anti-Cytomegalovirus Glycoprotein H Mouse monoclonal antibody [clone: 10B2669]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9100-22EEA 5940 DKK
USBIC9100-22E
Anti-Cytomegalovirus Glycoprotein H Mouse monoclonal antibody [clone: 10B2669]
Antistoffer
Recommended dilution: ELISA: 1:20-1:200; Immunofluorescence: 1:10-1:50; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Cytomegalovirus Glycoprotein H
Clonality: Monoclonal
Clone: 10B2669
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR