Anti-Cytomegalovirus, Glycoprotein B Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9100-21FEA 3040 DKK
USBIC9100-21F
Anti-Cytomegalovirus, Glycoprotein B Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:200-1:1000; Immunochemistry: 1:20-1:200; Western Blot: 1:20-1:200; Immunofluorescence: 1:2000

Type: Primary
Antigen: Cytomegalovirus, Glycoprotein B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR