Anti-Cytomegalovirus, Glycoprotein B Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9100-21CEA 2800 DKK
USBIC9100-21C
Anti-Cytomegalovirus, Glycoprotein B Goat polyclonal antibody
Antistoffer
Recommended dilution: Immunofluorescence: 1:2000

Type: Primary
Antigen: Cytomegalovirus, Glycoprotein B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR