Anti-Cytomegalovirus, 65kD Late Major Mouse monoclonal antibody [clone: 10B2656]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9100-15EEA 4880 DKK
USBIC9100-15E
Anti-Cytomegalovirus, 65kD Late Major Mouse monoclonal antibody [clone: 10B2656]
Antistoffer
Recommended dilution: Immunofluorescence: 1:10-1:50; Western Blot: 1:10-1:50

Type: Primary
Antigen: Cytomegalovirus, 65kD Late Major
Clonality: Monoclonal
Clone: 10B2656
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR